Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206980.1 | NP_996863.1 | 3 - 4 | 465 | 85 | |
XM_015284823.2 | XP_015140309.1 | 3 - 4 | 513 | 85 | ||
GenBank mRNA | AY371201.1 | - | 3 - 4 | 465 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |