Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTS2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206989.2 | NP_996872.1 | 4 - 5 | 413 | 68 | |
GenBank mRNA | AY566611.1 | - | 4 - 5 | 413 | 68 | |
CR406384.1 | - | 4 - 5 | 407 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |