Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THRSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213577.2 | NP_998742.1 | 1 - 2 | 458 | 133 | |
GenBank mRNA | AY568628.1 | - | 1 - 2 | 449 | 133 | |
AY568629.1 | - | 1 - 2 | 440 | 133 | ||
BX932096.2 | - | 1 - 2 | 446 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |