Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUCKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213581.2 | NP_998746.1 | 482 | 103 | ||
GenBank mRNA | AB049826.1 | - | 482 | 103 | ||
AJ720297.1 | - | 482 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus |