Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MESDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030551.2 | NP_001025722.2 | 2 - 3 | 424 | 75 | |
GenBank mRNA | AJ720080.1 | - | 2 - 3 | 424 | 75 | |
CR353965.1 | - | 2 - 3 | 341 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |