Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030552.1 | NP_001025723.1 | 8 - 9 | 1130 | 122 | |
XM_015292092.2 | XP_015147578.1 | 7 - 8 | 1050 | 122 | ||
GenBank mRNA | AJ719607.1 | - | 8 - 9 | 1130 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |