Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030589.1 | NP_001025760.1 | 4 - 5 | 452 | 96 | |
XM_015292829.1 | XP_015148315.1 | 4 - 5 | 470 | 96 | ||
XM_015292830.1 | XP_015148316.1 | 4 - 5 | 470 | 96 | ||
XM_015292831.2 | XP_015148317.1 | 4 - 5 | 483 | 96 | ||
XM_015292832.1 | XP_015148318.1 | 4 - 5 | 470 | 96 | ||
XM_015292833.1 | XP_015148319.1 | 4 - 5 | 470 | 96 | ||
XM_015292834.1 | XP_015148320.1 | 4 - 5 | 470 | 96 | ||
XM_015292835.1 | XP_015148321.1 | 4 - 5 | 470 | 96 | ||
XM_015292836.1 | XP_015148322.1 | 4 - 5 | 470 | 96 | ||
XM_015292837.2 | XP_015148323.1 | 4 - 5 | 485 | 96 | ||
GenBank mRNA | AJ719549.1 | - | 4 - 5 | 452 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |