Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030800.1 | NP_001025971.1 | 15 - 16 | 1863 | 86 | |
GenBank mRNA | AJ851409.1 | - | 15 - 16 | 1863 | 86 | |
MH119574.1 | - | 15 - 16 | 1750 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |