Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030801.2 | NP_001025972.2 | 6 - 7 | 696 | 112 | |
XM_015301559.2 | XP_015157045.1 | 6 - 7 | 683 | 112 | ||
GenBank mRNA | AB188299.2 | - | 6 - 7 | 696 | 112 | |
AJ719432.1 | - | 6 - 7 | 661 | 113 | ||
CR406923.1 | - | 4 - 5 | 350 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |