Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030820.1 | NP_001025991.1 | 7 - 8 | 735 | 90 | |
XM_015275343.2 | XP_015130829.1 | 9 - 10 | 995 | 90 | ||
XM_015275348.2 | XP_015130834.1 | 8 - 9 | 871 | 90 | ||
XM_015275351.2 | XP_015130837.1 | 8 - 9 | 935 | 90 | ||
GenBank mRNA | AJ851545.1 | - | 7 - 8 | 735 | 90 | |
CR352953.1 | - | 8 - 9 | 638 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |