Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030823.2 | NP_001025994.2 | 11 - 12 | 778 | 103 | |
XM_015275411.2 | XP_015130897.1 | 11 - 12 | 723 | 103 | ||
GenBank mRNA | AJ720706.1 | - | 10 - 11 | 853 | 103 | |
BU299206.1 | - | 10 - 11 | 662 | 103 | ||
BU471834.1 | - | 1 - 2 | 104 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |