Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030823.2 | NP_001025994.2 | 10 - 11 | 690 | 130 | |
XM_015275411.2 | XP_015130897.1 | 10 - 11 | 635 | 130 | ||
GenBank mRNA | AJ720706.1 | - | 9 - 10 | 765 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |