Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030998.1 | NP_001026169.1 | 2 - 3 | 294 | 145 | |
XM_015275876.2 | XP_015131362.1 | 2 - 3 | 462 | 145 | ||
GenBank mRNA | AJ720528.1 | - | 2 - 3 | 294 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |