Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031043.1 | NP_001026214.1 | 7 - 8 | 1394 | 146 | |
XM_025148668.1 | XP_025004436.1 | 8 - 9 | 5913 | 146 | ||
GenBank mRNA | AJ720620.1 | - | 7 - 8 | 1394 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |