Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031092.1 | NP_001026263.1 | 1 - 2 | 167 | 72 | |
GenBank mRNA | AJ719367.1 | - | 1 - 2 | 167 | 72 | |
BX950470.1 | - | 1 - 2 | 117 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |