Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031167.1 | NP_001026338.1 | 1 - 2 | 654 | 83 | |
XM_025149713.1 | XP_025005481.1 | 2 - 3 | 484 | 83 | ||
XM_025149714.1 | XP_025005482.1 | 2 - 3 | 484 | 83 | ||
XM_025149715.1 | XP_025005483.1 | 2 - 3 | 484 | 83 | ||
GenBank mRNA | AJ851513.1 | - | 1 - 2 | 654 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |