Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031167.1 | NP_001026338.1 | 3 - 4 | 992 | 138 | |
XM_025149713.1 | XP_025005481.1 | 4 - 5 | 822 | 138 | ||
GenBank mRNA | AJ851513.1 | - | 3 - 4 | 992 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |