Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031257.1 | NP_001026428.1 | 1 - 2 | 258 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |