Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031268.1 | NP_001026439.1 | 7 - 8 | 1225 | 134 | |
XM_015290281.2 | XP_015145767.1 | 8 - 9 | 1350 | 134 | ||
GenBank mRNA | AJ720888.1 | - | 7 - 8 | 1225 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |