Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031443.2 | NP_001026614.1 | 5 - 6 | 528 | 141 | |
GenBank mRNA | AJ719957.1 | - | 5 - 6 | 529 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |