Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031503.1 | NP_001026674.1 | 5 - 6 | 432 | 103 | |
GenBank mRNA | AJ719649.1 | - | 5 - 6 | 432 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |