Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031595.1 | NP_001026766.1 | 4 - 5 | 495 | 92 | |
GenBank mRNA | AJ719672.1 | - | 4 - 5 | 495 | 92 | |
BX934361.2 | - | 4 - 5 | 453 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |