Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270716.1 | NP_001257645.1 | 1 - 2 | 103 | 133 | |
GenBank mRNA | AJ721085.1 | - | 1 - 2 | 111 | 133 | |
BU296761.1 | - | 1 - 2 | 113 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |