Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030670.1 | NP_001025841.1 | 17 - 18 | 973 | 79 | |
NM_001031810.1 | NP_001026980.1 | 17 - 18 | 957 | 79 | ||
GenBank mRNA | M27665.1 | - | 2 - 3 | 93 | 79 | |
M27667.1 | - | 4 - 5 | 93 | 79 | ||
M61860.1 | - | 17 - 18 | 957 | 79 | ||
M61863.1 | - | 2090 | 79 | |||
M61864.1 | - | 17 - 18 | 973 | 79 | ||
MH156615.1 | - | 16 - 17 | 984 | 79 | ||
MH156616.1 | - | 16 - 17 | 984 | 79 | ||
MH156617.1 | - | 17 - 18 | 984 | 79 | ||
MH156618.1 | - | 16 - 17 | 1094 | 79 | ||
MH156619.1 | - | 17 - 18 | 984 | 79 | ||
MH156620.1 | - | 17 - 18 | 984 | 79 | ||
MH156621.1 | - | 15 - 16 | 1028 | 79 | ||
MH156623.1 | - | 17 - 18 | 984 | 79 | ||
MH156641.1 | - | 16 - 17 | 984 | 79 |
Gene Symbol: LOC425214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025155969.1 | XP_025011737.1 | 10 - 11 | 359 | 79 | |
XM_025155970.1 | XP_025011738.1 | 10 - 11 | 361 | 79 | ||
XM_025155971.1 | XP_025011739.1 | 10 - 11 | 355 | 79 | ||
XM_025155972.1 | XP_025011740.1 | 10 - 11 | 363 | 79 | ||
XM_025155973.1 | XP_025011741.1 | 10 - 11 | 364 | 79 | ||
XM_025155974.1 | XP_025011742.1 | 10 - 11 | 361 | 79 | ||
XM_025155975.1 | XP_025011743.1 | 10 - 11 | 363 | 79 | ||
XM_025155976.1 | XP_025011744.1 | 10 - 11 | 361 | 79 | ||
XM_025155977.1 | XP_025011745.1 | 10 - 11 | 359 | 79 | ||
XM_025155978.1 | XP_025011746.1 | 10 - 11 | 362 | 79 | ||
XM_025155979.1 | XP_025011747.1 | 10 - 11 | 362 | 79 | ||
XM_025155980.1 | XP_025011748.1 | 10 - 11 | 363 | 79 | ||
XM_025155981.1 | XP_025011749.1 | 10 - 11 | 360 | 79 | ||
XM_025155982.1 | XP_025011750.1 | 10 - 11 | 363 | 79 | ||
XM_025155983.1 | XP_025011751.1 | 10 - 11 | 362 | 79 | ||
XM_025155984.1 | XP_025011752.1 | 10 - 11 | 363 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |