Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030681.1 | NP_001025852.1 | 5 - 6 | 1470 | 124 | |
GenBank mRNA | AJ720914.1 | - | 5 - 6 | 1470 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |