Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030776.2 | NP_001025947.1 | 7 - 8 | 715 | 153 | |
XM_015293367.2 | XP_015148853.1 | 7 - 8 | 770 | 153 | ||
XM_015293370.2 | XP_015148856.1 | 7 - 8 | 739 | 153 | ||
XM_015293372.2 | XP_015148858.1 | 6 - 7 | 595 | 153 | ||
GenBank mRNA | AJ851464.1 | - | 7 - 8 | 716 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |