Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030894.2 | NP_001026065.1 | 4 - 5 | 517 | 67 | |
XM_015297648.2 | XP_015153134.1 | 5 - 6 | 664 | 67 | ||
GenBank mRNA | AJ719961.1 | - | 4 - 5 | 518 | 67 | |
BX930386.1 | - | 4 - 5 | 500 | 67 | ||
BX930954.1 | - | 5 - 6 | 594 | 67 | ||
BX932588.1 | - | 4 - 5 | 501 | 67 | ||
CR389031.1 | - | 4 - 5 | 498 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |