Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030949.2 | NP_001026120.2 | 3 - 4 | 339 | 118 | |
XM_015283013.2 | XP_015138499.1 | 4 - 5 | 500 | 118 | ||
XM_025147117.1 | XP_025002885.1 | 3 - 4 | 382 | 118 | ||
XM_025147118.1 | XP_025002886.1 | 4 - 5 | 546 | 118 | ||
XM_025147119.1 | XP_025002887.1 | 3 - 4 | 339 | 118 | ||
GenBank mRNA | AJ453193.1 | - | 3 - 4 | 340 | 118 | |
AJ721055.1 | - | 3 - 4 | 340 | 118 | ||
BU132419.1 | - | 3 - 4 | 270 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |