Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031341.1 | NP_001026512.1 | 13 - 14 | 1835 | 118 | |
XM_025143981.1 | XP_024999749.1 | 12 - 13 | 1608 | 118 | ||
GenBank mRNA | AJ720268.1 | - | 13 - 14 | 1835 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |