Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031343.1 | NP_001026514.1 | 4 - 5 | 671 | 95 | |
XM_025147313.1 | XP_025003081.1 | 4 - 5 | 899 | 95 | ||
GenBank mRNA | AJ720155.1 | - | 4 - 5 | 671 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |