Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031370.2 | NP_001026541.1 | 3 - 4 | 862 | 95 | |
GenBank mRNA | AJ851535.1 | - | 3 - 4 | 863 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |