Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031371.1 | NP_001026542.1 | 3 - 4 | 238 | 93 | |
GenBank mRNA | AJ719528.1 | - | 3 - 4 | 238 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |