Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031371.1 | NP_001026542.1 | 5 - 6 | 492 | 72 | |
GenBank mRNA | AJ719528.1 | - | 5 - 6 | 492 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |