Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031408.1 | NP_001026579.1 | 3 - 4 | 158 | 91 | |
XM_015300078.2 | XP_015155564.1 | 3 - 4 | 321 | 91 | ||
GenBank mRNA | AJ720270.1 | - | 3 - 4 | 158 | 91 | |
CR389795.1 | - | 3 - 4 | 334 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |