Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033850.1 | NP_001029022.1 | 5 - 6 | 500 | 91 | |
NM_001193588.1 | NP_001180517.1 | 7 - 8 | 647 | 91 | ||
NM_001193589.1 | NP_001180518.1 | 6 - 7 | 572 | 91 | ||
XM_025150837.1 | XP_025006605.1 | 7 - 8 | 830 | 91 | ||
GenBank mRNA | AB234613.1 | - | 7 - 8 | 647 | 91 | |
AB234614.1 | - | 6 - 7 | 572 | 91 | ||
AB234615.1 | - | 5 - 6 | 500 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |