Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039320.1 | NP_001034409.1 | 1 - 2 | 113 | 69 | |
GenBank mRNA | AJ719316.1 | - | 1 - 2 | 113 | 69 | |
KX395903.1 | - | 1 - 2 | 67 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |