Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039320.1 | NP_001034409.1 | 5 - 6 | 578 | 68 | |
GenBank mRNA | AJ719316.1 | - | 5 - 6 | 578 | 68 | |
CR386558.1 | - | 1 - 2 | 104 | 68 | ||
KX395903.1 | - | 5 - 6 | 532 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |