Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030797.2 | NP_001025968.1 | 1 - 2 | 273 | 79 | |
GenBank mRNA | EF408909.1 | - | 1 - 2 | 273 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |