Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037837.2 | NP_001032926.2 | 2 - 3 | 293 | 87 | |
GenBank mRNA | DQ275160.1 | - | 2 - 3 | 259 | 87 | |
DQ643391.1 | - | 2 - 3 | 293 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |