Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044651.1 | NP_001038116.1 | 7 - 8 | 776 | 111 | |
GenBank mRNA | DQ358914.1 | - | 7 - 8 | 776 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |