Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045837.1 | NP_001039302.1 | 13 - 14 | 1869 | 143 | |
XM_015284957.1 | XP_015140443.1 | 15 - 16 | 2038 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |