Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048076.2 | NP_001041541.2 | 2 - 3 | 681 | 134 | |
GenBank mRNA | BU132443.1 | - | 2 - 3 | 103 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |