Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080872.1 | NP_001074341.1 | 2 - 3 | 2705 | 124 | |
XM_015296420.2 | XP_015151906.1 | 2 - 3 | 2679 | 124 | ||
XM_015296421.2 | XP_015151907.1 | 2 - 3 | 2621 | 124 | ||
XM_025142271.1 | XP_024998039.1 | 2 - 3 | 2610 | 124 | ||
GenBank mRNA | AJ851732.1 | - | 2 - 3 | 2705 | 124 | |
AY342354.1 | - | 2 - 3 | 2696 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |