Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097526.1 | NP_001090995.1 | 5 - 6 | 385 | 102 | |
XM_015295706.1 | XP_015151192.1 | 6 - 7 | 646 | 102 | ||
GenBank mRNA | CR353595.1 | - | 6 - 7 | 650 | 102 | |
EF463065.1 | - | 5 - 6 | 385 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |