Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097637.1 | NP_001091106.1 | 3 - 4 | 214 | 104 | |
GenBank mRNA | BX934105.1 | - | 3 - 4 | 233 | 104 | |
BX935019.2 | - | 3 - 4 | 214 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |