Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM7.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099354.1 | NP_001092824.1 | 4 - 5 | 837 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |