Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105315.1 | NP_001098785.1 | 6 - 7 | 639 | 105 | |
NM_205130.1 | NP_990461.1 | 7 - 8 | 928 | 105 | ||
GenBank mRNA | D13516.1 | - | 7 - 8 | 928 | 105 | |
DQ870920.1 | - | 7 - 8 | 819 | 105 | ||
DQ870921.1 | - | 5 - 6 | 553 | 105 | ||
DQ870922.1 | - | 6 - 7 | 639 | 105 | ||
DQ870923.1 | - | 4 - 5 | 373 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |