Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109692.1 | NP_001103162.1 | 2 - 3 | 219 | 96 | |
XM_015282328.2 | XP_015137814.1 | 2 - 3 | 203 | 96 | ||
GenBank mRNA | AJ449410.1 | - | 2 - 3 | 221 | 96 | |
CR386184.1 | - | 2 - 3 | 190 | 96 | ||
CR387537.1 | - | 2 - 3 | 149 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |