Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115079.1 | NP_001108551.1 | 2 - 3 | 247 | 96 | |
GenBank mRNA | AB518061.1 | - | 2 - 3 | 285 | 96 | |
AB518062.1 | - | 3 - 4 | 371 | 96 | ||
AB518063.1 | - | 3 - 4 | 409 | 96 | ||
AB518064.1 | - | 4 - 5 | 521 | 96 | ||
AB518065.1 | - | 2 - 3 | 651 | 96 | ||
AB518066.1 | - | 2 - 3 | 337 | 96 | ||
AB518067.1 | - | 2 - 3 | 442 | 96 | ||
EU370215.1 | - | 1 - 2 | 198 | 96 | ||
EU370216.1 | - | 2 - 3 | 247 | 96 | ||
EU370217.1 | - | 2 - 3 | 231 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |