Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204132.1 | NP_989463.1 | 2 - 3 | 306 | 91 | |
GenBank mRNA | AY037891.1 | - | 2 - 3 | 306 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |